missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TYKi Antibody (CL14479), Novus Biologicals™
Mouse Monoclonal Antibody
€ 433.00 - € 629.00
Specifications
| Antigen | TYKi |
|---|---|
| Clone | CL14479 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043819
|
Novus Biologicals™
NBP3-26857-100ul |
100 μL |
€ 629.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043093
|
Novus Biologicals™
NBP3-26857-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TYKi Monoclonal antibody specifically detects TYKi in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| TYKi | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| PBS, pH 7.2, 40% glycerol | |
| 129607 | |
| IgG1 | |
| Protein A purified |
| CL14479 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
| This antibody was developed against a Synthetic Peptide of Q5EBM0. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title