missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38563
This item is not returnable.
View return policy
Description
TYW1B Polyclonal specifically detects TYW1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TYW1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9NV66 | |
| TYW1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RVMSRGEGDCDVVKSKHGSIEANFRAWKTKFISQLQALQKGERKKSCGGHCKKGKCESHQHGSEEREEGSQEQDELHHR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| LINC00069, Long Intergenic Non-Protein Coding RNA 69, NCRNA00069, Non-Protein Coding RNA 69, Radical S-Adenosyl Methionine And Flavodoxin Domain-Containing Protein 2, radical S-adenosyl methionine and flavodoxin domains 1, RSAFD2, S-Adenosyl-L-Methionine-Dependent TRNA 4-Demethylwyosine Synthase, TRNA Wybutosine-Synthesizing Protein 1 Homolog B TRNA-YW Synthesizing Protein 1 Homolog B, TRNA-YW Synthesizing Protein 1 Homolog B (Non-Protein Coding), TRNA-YW Synthesizing Protein 1 Homolog B (S. Cerevisiae) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55253 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction