Learn More
uvex U-Chem 3100 Chemical Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
Description
- Nitrile butadiene rubber (NBR) chemical protection gloves
- An additional coating enhances grip capabilities when handling wet/oily objects
- Ideal for a wide variety of applications that require mechanical protection
Specifications
Specifications
| Certifications/Compliance | EN 388 (4 1 2 1 X), EN ISO 374-1:2016 / Type A (A J K L M O) |
| Color | Black |
| Latex Free | Yes |
| Material | Cotton, NBR |
| Product Type | Chemical-resistant Gloves |
| Size | 11 |