Learn More
Uvex™ U-Chem 3200 Chemical Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
Description
- NBR-coated chemical protection gloves with nylon weave supporting material
- Slim-fit design easily stretches and naturally moves with the hand for increased dexterity
- Additional coating on the palm ensures a secure grip
- Resistant to grease, mineral oils, and various chemicals
- Ideal for handling oil, cooling lubricants, and grease
Specifications
Specifications
| Applications | Laboratory |
| Coating Material | NBR |
| Color | Blue, Black |
| Size | 8 |
| EN388 Standard | 3131X |
| EN ISO 374-1 | Type A |
| EN ISO 374-1 CHEMICAL LETTER CODES | JKLMOT |
| For Use With (Application) | Handling Oil, Cooling Lubricants, and Chemicals |
| Gender | Unisex |
| Glove Cuff Style | Gauntlet |
| Show More |