Learn More
uvex U-Chem 3500 Safety Glove
Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
Description
- Chemical protection glove with gauntlet for handling a variety of different chemicals
- Fully coated with chloroprene and nitrile rubber (NBR)
- Cotton top material
- Excellent grip on warm and wet workpieces
- High resistance to acetone, cleaning agents, adhesives and solvents
- Very comfortable to wear thanks to seamless cotton liner
Specifications
Specifications
| Certifications/Compliance | EN 388:2016 (3 1 2 1 X), EN 374: Type A (A C J K L M N O P S T), EN 407 (X 1 X X X X) |
| Color | Orange |
| Latex Free | Yes |
| Material | Cotton, NBR |
| Product Type | Chemical-resistant Gloves |
| Size | 11 |