Learn More
uvex U-Chem 3500 Safety Glove
Product Code. p-7228494 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
Product Code. 17797739
Brand: uvex 6018807
The Uvex U-chem 3500 chemical protection gloves are ideal for handling a variety of different chemicals.
  • Chemical protection glove with gauntlet for handling a variety of different chemicals
  • Fully coated with chloroprene and nitrile rubber (NBR)
  • Cotton top material
  • Excellent grip on warm and wet workpieces
  • High resistance to acetone, cleaning agents, adhesives and solvents
  • Very comfortable to wear thanks to seamless cotton liner
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388:2016 (3 1 2 1 X), EN 374: Type A (A C J K L M N O P S T), EN 407 (X 1 X X X X)
Color Orange
Latex Free Yes
Material Cotton, NBR
Product Type Chemical-resistant Gloves
Size 7