missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAF1/WDR48 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49269
This item is not returnable.
View return policy
Description
UAF1/WDR48 Polyclonal antibody specifically detects UAF1/WDR48 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| UAF1/WDR48 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| KIAA1449DKFZp686G1794, P80, UAF1, USP1 associated factor 1, USP1-associated factor 1, WD repeat domain 48, WD repeat endosomal protein, WD repeat-containing protein 48 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SDMLQVRKVMEHVYEKIINLDNESQTTSSSNNEKPGEQEKEEDIAVLAEEKIELLCQDQVLDPNMDLRTVKHFIWKSGGDLTLHYR | |
| 0.1 mL | |
| Cell Biology | |
| 57599 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction