Learn More
UBC12 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H8D2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67482-20UL
 View more versions of this product
Product Code. 18028795
  • € 145.00 / 20µL
Description
The UBC12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBC12. This antibody recognizes human, mouse, rat antigen. The UBC12 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
UBC12
1H8D2
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
hUbc12, NEDD8 carrier protein, NEDD8 conjugating enzyme Ubc12, NEDD8 protein ligase, UBC RS2, UBC12, UBE2M
20 μL
9040
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.