Learn More
UBC12 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H8D2, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | UBC12 |
|---|---|
| Clone | 1H8D2 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068315
|
Proteintech
CL488-67482-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028795
|
Proteintech
CL488-67482-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UBC12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBC12. This antibody recognizes human, mouse, rat antigen. The UBC12 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| UBC12 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 9040 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1H8D2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| hUbc12, NEDD8 carrier protein, NEDD8 conjugating enzyme Ubc12, NEDD8 protein ligase, UBC RS2, UBC12, UBE2M | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.