missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Mouse anti-Nonspecies specific, Clone: FK1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11039-25UG
This item is not returnable.
View return policy
Description
Ubiquitin Monoclonal specifically detects Ubiquitin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Ubiquitin | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:10000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Ubiquitin conjugated lysozyme (Uniprot: P0CG47) | |
| 25 μg | |
| Primary | |
| Non-species specific | |
| IgM |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| FK1 | |
| Unconjugated | |
| 10 mM phosphate buffer (pH 7.4), 0.15 M NaCl | |
| Mouse | |
| Gel filtration | |
| RUO | |
| 6233 | |
| Store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction