missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Mouse anti-Nonspecies specific, Clone: FK1, Novus Biologicals™
Mouse Monoclonal Antibody
€ 255.00 - € 571.00
Specifications
| Antigen | Ubiquitin |
|---|---|
| Clone | FK1 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:10000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18083866
|
Novus Biologicals
NBP3-11039-25UG |
25 μg |
€ 255.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074826
|
Novus Biologicals
NBP3-11039-100UG |
100 μg |
€ 571.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ubiquitin Monoclonal specifically detects Ubiquitin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Ubiquitin | |
| 1 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| 10 mM phosphate buffer (pH 7.4), 0.15 M NaCl | |
| 6233 | |
| IgM | |
| Gel filtration |
| FK1 | |
| Western Blot 1:10000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Mouse | |
| Non-species specific | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Ubiquitin conjugated lysozyme (Uniprot: P0CG47) | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title