missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP1/3 Antibody (SR1570), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22657-50ul
This item is not returnable.
View return policy
Description
UCP1/3 Monoclonal antibody specifically detects UCP1/3 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| UCP1/3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| A synthesized peptide derived from human UCP1/3 (Uniprot #: P12242, P56501) | |
| 50 μg | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| SR1570 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:50 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction