missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1, p Ser556 Antibody (SR1119), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22612-50ul
This item is not returnable.
View return policy
Description
ULK1 Monoclonal antibody specifically detects ULK1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ULK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1119 | |
| Western Blot 1:500-1:2000 | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| A synthesized peptide derived from human phospho-ULK1 [Ser556] (Uniprot #: O75385) | |
| 50 μg | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| 8408 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction