missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1, p Ser556 Antibody (SR1119), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | ULK1 |
|---|---|
| Clone | SR1119 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685204
|
Novus Biologicals
NBP3-22612-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613694
|
Novus Biologicals
NBP3-22612-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULK1 Monoclonal antibody specifically detects ULK1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ULK1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| A synthesized peptide derived from human phospho-ULK1 [Ser556] (Uniprot #: O75385) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1119 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8408 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title