Learn More
uvex Unilite Thermo Safety Gloves
Product Code. p-9618006 Shop All Uvex Products
Click to view available options
Size:
7
11
Unit Size:
10 pairs
Each
Product Code. 17290461
Brand: uvex 6059307
Only null left
Add to basket
Add to basket
Two-layer material composition features acrylic and new wool to ensure a pleasant hand climate and enhanced user comfort. Polymer coating provides mechanical abrasion resistance, and remains flexible even at low temperatures.
  • Gloves feature two-layer material composition of acrylic and new wool
  • Polymer coating on the palm remains flexible even at low temperatures
  • Coating provides mechanical abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp areas
  • Provides thermal insulation when in direct contact with cold objects
  • Safe for use when handling food
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 511: 010
Applications Industrial
Coating Material Polymer
Color Black
Exterior Finish Smooth
Hand Ambidextrous
Material Polyamide, Elastane
Product Type Cold-resistant Gloves
Size 7
EN388 Standard 3131X
For Use With (Application) Construction, Forklift Operation, and Refrigeration
Gender Unisex
Glove Cuff Style Knit
Liner Material Acrylic, Wool
Series Unilite Thermo