Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 10773863 Shop All Fisherbrand Products
Click to view available options
pH Range:
0.5 to 5
0.5 to 5.5
1 to 11
1 to 14
12 to 14
4 to 7
5 to 9
6.4 to 8
6.5 to 10
9 to 13
Format:
Refill Pack
Roll
Tube
Length (Metric):
5 m
50 mm
Quantity:
1 Roll
10 Pack
3 Pack
4 Rolls
Unit Size:
Each
Pack of 3
Pack of 4
Product Code. 10773863
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Gives rough knowledge of pH.
  • Filter paper soaked with one or several colored indicators of pH
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range 1 to 11
Format Refill Pack
Includes Twist-lock dispensers and color scale
Quantity 3 Pack
Type Grade 911
Length (Metric) 5 m
Width (Metric) 10 mm