missing translation for 'onlineSavingsMsg'
Learn More

URIDINE-5'-DIPHOSPHATE, MP Biomedicals

Product Code. 11398819 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11398819

Brand: MP Biomedicals 0210120591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Disodium Salt

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-98-0
Molecular Formula C9H14N2O12P2
Molecular Weight (g/mol) 404.16
InChI Key XCCTYIAWTASOJW-UHFFFAOYNA-N
Synonym uridine 5'-diphosphate, uridine diphosphate, uridine 5'-trihydrogen diphosphate, udp, uridine-5'-diphosphate, unii-5g0f599a1y, 5'-udp, uridine 5'-pyrophosphate, uridine 5'-pyrophosphoric acid, uridine-diphosphate
PubChem CID 6031
ChEBI CHEBI:17659

Specifications

Quantity 1 g
Formula Weight 448.17 (Anhydrous)
Chemical Name or Material URIDINE-5'-DIPHOSPHATE, MP Biomedicals
compliance-icons compliance-icons
Product Identifier
  • URIDINE-5`-DIPHOSPHATE DISODIUM SALT
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H371-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Uridine-5'-diphosphate Disodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.