missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP11L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35848-20ul
This item is not returnable.
View return policy
Description
UTP11L Polyclonal antibody specifically detects UTP11L in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| UTP11L | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-240 of human UTP11L (NP_057121.2).,, Sequence:, VAEAKKIERLKSELHLLDFQGKQQNKHVFFFDTKKEVEQFDVATHLQTAPELVDRVFNRPRIETLQKEKVKGVTNQTGLKRIAKERQKQYNCLTQRIEREKKLFVIAQKIQTRKDLMDKTQKVKVKKETVN | |
| 20 μL | |
| Cell Cycle and Replication, Cytokine Research | |
| 51118 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction