missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93662-0.1ml
This item is not returnable.
View return policy
Description
UTP3 Polyclonal antibody specifically detects UTP3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UTP3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-479 of human UTP3 (NP_065101.1). KLSVVDQKLSSEIRHLLTLKDDAVKKELIPKAKSTKPKPKSVSKTSAAACAVTDLSDDSDFDEKAKLKYYKEIEDRQKLKRKKEENSTEEQALEDQNAKRAITYQIAKNRGLTPRRKKIDRNPRVKHREKFRRAKIRRRGQVREVRKEEQRYSGELSGIRAGVKKSIKLK | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 57050 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction