missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | UTP3 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613461
|
Novus Biologicals
NBP2-93662-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691461
|
Novus Biologicals
NBP2-93662-0.1ml |
0.1 mL |
€ 470.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UTP3 Polyclonal antibody specifically detects UTP3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| UTP3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers | |
| PBS (pH 7.3), 50% glycerol | |
| 57050 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-479 of human UTP3 (NP_065101.1). KLSVVDQKLSSEIRHLLTLKDDAVKKELIPKAKSTKPKPKSVSKTSAAACAVTDLSDDSDFDEKAKLKYYKEIEDRQKLKRKKEENSTEEQALEDQNAKRAITYQIAKNRGLTPRRKKIDRNPRVKHREKFRRAKIRRRGQVREVRKEEQRYSGELSGIRAGVKKSIKLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title