Learn More
uvex Profapren CF33 Chemical Protecion Gloves
Product Code. p-7160263 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
Each
Product Code. 15336922
Brand: uvex 6011901
High-quality unsupported chloroprene safety gloves for protection against a broad spectrum of chemicals
  • Silicone-free safety gloves with excellent properties for protection against chemical and mechanical risks
  • Good combination of flexibility and strength
  • Resistant to many chemicals and solvents
  • EN 388:2016 (3131X)
  • EN ISO 374-1: 2016 / Type A (AKLMNO)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 388:2016 (3 1 3 1 X), 374-1:2016 /Type A (A K L M N O)
    Coating Material Polychloroprene
    Color Blue
    Latex Free Yes
    Material Cotton
    Product Type Chemical-resistant Gloves
    Size 7
    Thickness (Metric) 0.75 mm
    For Use With (Application) Chemical industry, painting/coating, metal work (cleaning)
    Model CF33
    Palm Type Napped
    Length (Metric) 33 cm