Learn More
Valeraldehyde (Pentanal), TRC
Product Code. 30009746
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009746 250 mg 250mg
30136535 1 g 1g
30005872 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009746 Supplier Toronto Research Chemicals Supplier No. TRCV091295250MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-62-3
Molecular Formula C5 H10 O
Molecular Weight (g/mol) 86.13
Synonym Valeraldehyde, Pentanal, Valeraldehyde (8CI), NSC 35404, Pentylaldehyde, Valeral, Valeric acid aldehyde, Valeric aldehyde, n-Pentanal, n-Valeraldehyde
IUPAC Name pentanal
SMILES CCCCC=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3
Formula Weight 86.0732
Chemical Name or Material n-Valeraldehyde