Learn More
Valerophenone p-toluenesulfonylhydrazone, 97%, Thermo Scientific™
Product Code. 15420158
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420158 5 g 5g
15430158 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15420158 Supplier Thermo Scientific Alfa Aesar Supplier No. H53392.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69015-74-3
Molecular Formula C18H22N2O2S
Molecular Weight (g/mol) 330.446
MDL Number MFCD00159338
InChI Key VTEOESQGMPJHGP-VHEBQXMUSA-N
Synonym valerophenone p-toluenesulfonylhydrazone, 4-methyl-n'-1e-1-phenylpentylidene benzenesulfonohydrazide, valerophenone tosylhydrazone, mixture of isomers, 4-methyl-n-e-1-phenylpentylideneamino benzenesulfonamide, 4-methyl-n'-1z-1-phenylpentylidene benzenesulfonohydrazide, benzenesulfonic acid, 4-methyl-, n'-e-1-phenylpentylidene hydrazide
PubChem CID 6374697
IUPAC Name 4-methyl-N-[(E)-1-phenylpentylideneamino]benzenesulfonamide
SMILES CCCCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)C2=CC=CC=C2
Specifications
Melting Point 128°C to 136°C
Quantity 5 g
Formula Weight 330.44
Percent Purity 97%
Chemical Name or Material Valerophenone p-toluenesulfonylhydrazone