missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP1 Monoclonal Antibody (K40030_15G9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555791
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: PPGPPPNMTS NRRLQQTQAQ VEEVVDIIRV NVDKVLERDQ KLSELDDRAD ALQAGASQFE SSAAKLKRKY
Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
Specifications
| VAMP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P23763 | |
| Vamp1 | |
| Recombinant human VAMP1 (immunogen range is 21-90). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K40030_15G9 | |
| Unconjugated | |
| Vamp1 | |
| DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1) | |
| Mouse | |
| Protein A | |
| RUO | |
| 6843 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction