missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP1 Monoclonal Antibody (K40030_15G9), Invitrogen™
Mouse Monoclonal Antibody
€ 438.00 - € 658.00
Specifications
| Antigen | VAMP1 |
|---|---|
| Clone | K40030_15G9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30194173
|
Invitrogen™
MA556171 |
200 μL |
€ 658.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30207992
|
Invitrogen™
MA555791 |
50 μL |
€ 438.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: PPGPPPNMTS NRRLQQTQAQ VEEVVDIIRV NVDKVLERDQ KLSELDDRAD ALQAGASQFE SSAAKLKRKY
VAMP1 Monoclonal antibody specifically detects VAMP1 in Human samples. It is validated for Western BlotSpecifications
| VAMP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1) | |
| Recombinant human VAMP1 (immunogen range is 21-90). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Vamp1 |
| K40030_15G9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23763 | |
| 6843 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title