Learn More
B Braun™ Vasco™OP Sensitive Sterile Glove
Click to view available options
Size:
6
7.5
8
Unit Size:
1 pair
Pack of 40
Description
- Medical surgical glove according to EN 455
- Protective gloves according to PSA 89/686 / EEC, EN 420, EN 374
- Easier working due to fully anatomical shape
- Very good grip thanks to micro-rough surface
- Excellent wearing comfort and excellent fit
- Perfect tactile sensitivity and optimal safety
- AQL: 0.65
Specifications
Specifications
| Cut Style | Rolled cuff |
| Color | Tan |
| Latex Free | No |
| Material | Latex |
| Powdered | Powder Free |
| Size | 8 |
| Sterility | Sterile |
| Glove Cuff Color | Tan |