Learn More
B Braun™ Vasco™OP Sensitive Sterile Glove
Product Code. 10461761
Change view
Click to view available options
Size:
6
7.5
8
Unit Size:
1 pair
Pack of 40
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10461761 7.5 1 pair
10094940 6 1 pair
10527604 7.5 Pack of 40
10412365 8 Pack of 40
10219080 8 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Warning: Gloves contain natural latex, the allergic Reactions including anaphylactic reactions can trigger
Product Code. 10461761 Supplier B Braun™ Supplier No. 6081037
  • ONLINE EXCLUSIVE PRICE
    € 1.20
    € 1.18 / 1 pairs
    Save € 0.02
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Warning: Gloves contain natural latex, the allergic Reactions including anaphylactic reactions can trigger
Gain a better grip with micro-rough surface glove. B Braun™ Vasco™OP Sensitive Sterile Glove with inner coating is made of highly elastic natural latex. These ergonomically shaped surgical gloves are comfortable, provide good tactile sensation and feature rolled cuffs.  
  • Medical surgical glove according to EN 455
  • Protective gloves according to PSA 89/686 / EEC, EN 420, EN 374
  • Easier working due to fully anatomical shape
  • Very good grip thanks to micro-rough surface
  • Excellent wearing comfort and excellent fit
  • Perfect tactile sensitivity and optimal safety
  • AQL: 0.65
Specifications
Cut Style Rolled cuff
Color Tan
Latex Free No
Material Latex
Powdered Powder Free
Size 7.5
Sterility Sterile
Glove Cuff Color Tan