Learn More
VASP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-16890-100UL
 Afficher plus de versions de ce produit
Code produit. 18027655
  • € 461.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
VASP Polyclonal antibody specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
VASP
Polyclonal
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
vasodilator-stimulated phosphoprotein
A synthetic phosphorylated peptide around S239 of human VASP (NP_003361.1).
100 μg
Phospho Specific
7408
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS, 50% glycerol, pH7.3