missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 461.00
Specifications
| Antigen | VASP |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389853
|
Novus Biologicals
NBP3-16890-20UL |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18027655
|
Novus Biologicals
NBP3-16890-100UL |
100 μg |
€ 461.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VASP Polyclonal antibody specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VASP | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, 50% glycerol, pH7.3 | |
| 7408 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| vasodilator-stimulated phosphoprotein | |
| A synthetic phosphorylated peptide around S239 of human VASP (NP_003361.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title