missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Antibody (B-K9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47864B
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| Biotin | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Activated human umbilical vein endothelial cells (HUVEC) | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 7412 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| B-K9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction