missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Antibody (B-K9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 595.00 - € 643.00
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | B-K9 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687525
|
Novus Biologicals
NBP2-47864B-100ul |
100 μg |
€ 643.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692936
|
Novus Biologicals
NBP2-47864B |
0.1 mL |
€ 595.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| VCAM-1/CD106 | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| PBS | |
| 7412 | |
| IgG1 κ | |
| Protein A or G purified |
| B-K9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Activated human umbilical vein endothelial cells (HUVEC) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title