missing translation for 'onlineSavingsMsg'
Learn More

VCP Polyclonal Antibody, OriGene

Rabbit Polyclonal Antibody

Brand:  Origene Technologies TA890028S

Encompass_Preferred

 View more versions of this product

Product Code. 17539434

  • € 227.48 / 30µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VCP Polyclonal antibody specifically detects VCP in Human samples. It is validated for Western Blot
Specifications

Specifications

VCP
Polyclonal
Unconjugated
RUNX2
15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog
Recombinant protein of human RUNX2
30 μL
Primary
Human
Liquid
Western Blot
1.77 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q13950
Rabbit
Antigen affinity chromatography
RUO
860
-20° C, Avoid Freeze/Thaw Cycles
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.