missing translation for 'onlineSavingsMsg'
Learn More

VCP Polyclonal Antibody, OriGene

Rabbit Polyclonal Antibody

€ 227.48 - € 492.47

Specifications

Antigen VCP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17539434
Encompass_Preferred
View Documents
Origene Technologies
TA890028S
30 μL
€ 227.48
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17529434
Encompass_Preferred
View Documents
Origene Technologies
TA890028
100 μL
€ 492.47
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VCP Polyclonal antibody specifically detects VCP in Human samples. It is validated for Western Blot
Specifications

Specifications

VCP
Polyclonal
Liquid
RUO
Primary
Western Blot
Unconjugated
Rabbit
IgG
-20° C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.