missing translation for 'onlineSavingsMsg'
Learn More

VDAC2 Antibody, Novus Biologicals™

Chicken Polyclonal Antibody

Brand:  Novus Biologicals NB100-74560-0.025mg

 View more versions of this product

Product Code. 18587163

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

VDAC2 Polyclonal specifically detects VDAC2 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VDAC2
Polyclonal
Unconjugated
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
VDAC2
VDAC2 Protein
0.025 mg
Primary
This detects VDAC2 in mouse, rat, human, rabbit, and bovine samples.
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:500, ELISA 1:100 - 1:2000
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Chicken
Affinity Purified
RUO
7417
Human
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only