missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 195.00
Specifications
| Antigen | VDAC2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, ELISA 1:100 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18587163
|
Novus Biologicals
NB100-74560-0.025mg |
0.025 mg | |||||||
Description
VDAC2 Polyclonal specifically detects VDAC2 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA.Specifications
| VDAC2 | |
| Western Blot 1:500, ELISA 1:100 - 1:2000 | |
| Polyclonal | |
| Chicken | |
| Human | |
| FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2 | |
| VDAC2 | |
| IgY | |
| Affinity Purified | |
| This detects VDAC2 in mouse, rat, human, rabbit, and bovine samples. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| 7417 | |
| VDAC2 Protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title