Learn More
VDR/NR1I1/Vitamin D Receptor Antibody (3C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26255-100ul
 View more versions of this product
Product Code. 30039164
  • € 541.00 / 100µL
Description
VDR/NR1I1/Vitamin D Receptor Monoclonal antibody specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VDR/NR1I1/Vitamin D Receptor
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
3C7
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor
A synthesized peptide derived from Human VDR/NR1I1/Vitamin D Receptor [UniProt P11473]
100 μL
Signal Transduction
7421
Store at -20 to -70C. Avoid freeze-thaw cycles.