missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (3D5C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37420-0.025ml
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| VE-Cadherin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Mouse | |
| 87.5 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 3D5C7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CDH5 | |
| Purified recombinant fragment of human CDH5 (AA: 29-223 ) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction