missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (3D5C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 208.00 - € 564.00
Specifications
| Antigen | VE-Cadherin |
|---|---|
| Clone | 3D5C7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18132899
|
Novus Biologicals
NBP2-37420 |
0.1 mL |
€ 564.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640656
|
Novus Biologicals
NBP2-37420-0.025ml |
0.025 mL |
€ 208.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| VE-Cadherin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| CDH5 | |
| IgG1 | |
| Affinity Purified |
| 3D5C7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 1003 | |
| Purified recombinant fragment of human CDH5 (AA: 29-223 ) expressed in E. Coli. | |
| Primary | |
| 87.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title