missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin, p Tyr731 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54766-0.05MG
This item is not returnable.
View return policy
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| VE-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
| CDH5 | |
| VE-Cadherin (phospho Tyr731) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y (p)-G-Y) derived from Human VE-Cadherin. | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000 | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Rabbit | |
| 0.05 mg | |
| 1003 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction