missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin, p Tyr731 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 195.00 - € 696.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235340
|
Novus Biologicals
NBP2-54766 |
0.1 mg |
€ 696.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18291102
|
Novus Biologicals
NBP2-54766-0.05MG |
0.05 mg |
€ 442.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671508
|
Novus Biologicals
NBP2-54766-0.025mg |
0.025 mg |
€ 195.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot.Specifications
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Human, Mouse | |
| 1003 | |
| VE-Cadherin (phospho Tyr731) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y (p)-G-Y) derived from Human VE-Cadherin. | |
| Primary |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| CDH5 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title