missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin, p Tyr731 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54766-0.025mg
This item is not returnable.
View return policy
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| VE-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| CDH5 | |
| VE-Cadherin (phospho Tyr731) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y (p)-G-Y) derived from Human VE-Cadherin. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody detects endogenous level of VE-Cadherin only when phosphorylated at tyrosine 731. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000 | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Rabbit | |
| 130 kDa | |
| 0.025 mg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction