missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Antibody (VEGFA/7635R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20679-20ug
This item is not returnable.
View return policy
Description
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| VEGF | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant fragment (around aa1-200) of human VEGF protein (exact sequence is proprietary) | |
| 20 μg | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 7422 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| VEGFA/7635R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction