missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Antibody (VEGFA/7635R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 287.00 - € 519.00
Specifications
| Antigen | VEGF |
|---|---|
| Clone | VEGFA/7635R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670075
|
Novus Biologicals
NBP3-20679-100ug |
100 μg |
€ 519.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663334
|
Novus Biologicals
NBP3-20679-20ug |
20 μg |
€ 287.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| VEGF | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 10mM PBS with 0.05% BSA | |
| 7422 | |
| IgG1 κ | |
| Protein A or G purified |
| VEGFA/7635R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Recombinant fragment (around aa1-200) of human VEGF protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title