missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGI Mouse, Clone: VEGI/1283, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 9966-MSM1-P0
This item is not returnable.
View return policy
Specifications
| VEGI | |
| Secreted and Cell Surface | |
| VEGI/1283 | |
| Unconjugated | |
| NP_001191273 | |
| Mouse | |
| 22kDa | |
| Protein A/G Purified | |
| RUO | |
| 9966 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| TNFSF15 | |
| Recombinant human full-length VEGI fragment | |
| Endothelial cells. Colon, intestine, placenta, lung, liver, kidney, pancreas, spleen and prostate. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction