missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGI Mouse, Clone: VEGI/1283, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | VEGI |
|---|---|
| Clone | VEGI/1283 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15901318
|
EnquireBio
9966-MSM1-P0 |
20 μg | |||||||
|
15911318
|
EnquireBio
9966-MSM1-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| VEGI | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 9966 | |
| Recombinant human full-length VEGI fragment | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 22kDa |
| VEGI/1283 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001191273 | |
| TNFSF15 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Secreted and Cell Surface | |
| Endothelial cells. Colon, intestine, placenta, lung, liver, kidney, pancreas, spleen and prostate. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title