Learn More
Ansell™ VersaTouch™ 92-220
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
Description
- Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions.
- A thinner glove with enhanced fingertip flexibility and improved overall comfort.
- Textured fingertips provide good comfort and tactility for sensitive, precise handling.
- Latex-free: no risk of Type 1 allergies.
- Approved for food contact applications. Ideal for handling all fatty food.
Specifications
Specifications
| Exterior Finish | Fully Textured |
| Material | Nitrile |
| Size | 7 |
| Thickness (English) | 2.9 mil |
| Thickness (Metric) | 0.07 mm |
| Model | 922207 |
| Series | 92-220 |