missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Versican Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
€ 375.00 - € 532.35
Specifications
| Antigen | Versican |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18490772
|
Novus Biologicals
NBP1-85432-25ul |
25 μL |
€ 375.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18001964
|
Novus Biologicals
NBP1-85432 |
0.1 mL |
€ 563.00 € 532.35 / 0.10mL Save € 30.65 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Versican Polyclonal specifically detects Versican in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Versican | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Extracellular Matrix | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Chondroitin sulfate proteoglycan 2WGN, Chondroitin sulfate proteoglycan core protein 2, CSPG2DKFZp686K06110, ERVR, GHAP, Glial hyaluronate-binding protein, Large fibroblast proteoglycan, PG-MWGN1, versican, versican core protein, versican proteoglycan | |
| VCAN | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P13611 | |
| 1462 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SLSGKVSLPCHFSTMPTLPPSYNTSEFLRIKWSKIEVDKNGKDLKETTVLVAQNGNIKIGQDYKGRVSVPTHPEAVGDASLTVVKLLASDAGLYRCDVMYGIEDTQDTVSLTVDGVVFHYRAATSRYTLNFEAA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title