Learn More
Honeywell™ Vertigo Black PU C&G 1 Gloves
Product Code. p-7161091
Click to view available options
Size:
6
7
8
9
10
Unit Size:
1 pair
Product Code. 15806695
Brand: Honeywell™ 213225106
Only null left
Add to basket
Add to basket
Honeywell™ Vertigo Black PU C&G 1 Gloves for handling of fine products in a dry, slightly greasy or soiled environment.
  • 100% black polyamide knitted glove
  • Black polyurethane coating on palm and fingertips
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • The polyamide and its polyurethane coating provide a good abrasion and tear resistance
  • Non pilling, the polyamide fiber is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting environment
  • Its dark color makes it more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standard, EN 388-2016 Mechanical risks
Description 13 gauge
Coating Material Polyamide, Polyurethane
Color Black
Size 6
Length (Metric) 25 cm