Learn More
Honeywell™ Vertigo Gray PU 3 Gloves
Description
- White Spectra™/lycra™ knitted glove
- Gray polyurethane coating on palm and tips of fingers
- Elastic wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The cool Spectra™ touch gives an added sensation of comfort
- The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The glove's ventilated back optimises its breathability
- The Spectra®/lycra® thread provides outstanding properties against cuts
- The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
- Caution! Do not expose this glove to the heat
- Its white color enhances the glove's hygiene and cleanliness
- Its gray coating enables work in a slightly soiled environment
Specifica
Specifica
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards |
| Description | 13 gauge |
| Color | Gray, White |
| Material | Polyurethane, Lycra |
| Size | 8 |
| Length (Metric) | 23.5 cm |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?