Learn More
Honeywell™ Vertigo Gray PU 3 Gloves
Product Code. p-7159540
Click to view available options
Size:
8
9
Unit Size:
10 pairs
Product Code. 15625947
Brand: Honeywell™ 231876708
Only null left
Add to basket
Edge
Add to basket
Honeywell™ Vertigo Gray PU 3 Gloves for handling sharp parts in a dry or greasy environment.
  • White Spectra™/lycra™ knitted glove
  • Gray polyurethane coating on palm and tips of fingers
  • Elastic wrist
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The cool Spectra™ touch gives an added sensation of comfort
  • The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The glove's ventilated back optimises its breathability
  • The Spectra®/lycra® thread provides outstanding properties against cuts
  • The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
  • Caution! Do not expose this glove to the heat
  • Its white color enhances the glove's hygiene and cleanliness
  • Its gray coating enables work in a slightly soiled environment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards
Description 13 gauge
Color Gray, White
Material Polyurethane, Lycra
Size 8
Length (Metric) 23.5 cm